Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzyl alcohol, 98%

Catalog No. AAB2126014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2126014 25 g
AAB2126006 5 g
AAB2126022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2126014 Supplier Thermo Scientific Chemicals Supplier No. B2126014
Add to Cart

CAS: 877-65-6 | C11H16O | 164.248 g/mol

4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-65-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00004662
InChI Key FVEINXLJOJPHLH-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol
PubChem CID 13416
IUPAC Name (4-tert-butylphenyl)methanol
SMILES CC(C)(C)C1=CC=C(C=C1)CO

Specifications

Density 0.928
Boiling Point 139°C to 140°C (20 mmHg)
Flash Point 128°C (262°F)
Refractive Index 1.517
Quantity 25 g
Beilstein 1860665
Solubility Information Soluble in alcohol and insoluble in water.
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzyl alcohol
EINECSNumber 212-894-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.