Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. B176125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176125G 25 g
B17615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B176125G Supplier TCI America Supplier No. B176125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-65-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00004662
InChI Key FVEINXLJOJPHLH-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol
PubChem CID 13416
IUPAC Name (4-tert-butylphenyl)methanol
SMILES CC(C)(C)C1=CC=C(C=C1)CO

Specifications

Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 164.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.