Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyldiphenyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. b348425g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B348425G 25 g
B3484100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B348425G Supplier TCI America Supplier No. B348425G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85609-03-6
Molecular Formula C16H18S
Molecular Weight (g/mol) 242.38
MDL Number MFCD06200845
InChI Key KGRAEJDVHXHGSQ-UHFFFAOYSA-N
PubChem CID 11413822
IUPAC Name 1-tert-butyl-4-phenylsulfanylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)SC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 242.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butyldiphenyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.