Learn More
4-tert-Butylphenol, 99%
Description
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.
Solubility
Soluble in water. [8.7 g/L (20°C)]
Notes
Store at room temperature. Keep away from oxidizing agents and acids.
Specifications
Specifications
| Melting Point | 97°C to 101°C |
| Density | 0.908 |
| Boiling Point | 236°C to 238°C |
| Flash Point | 113°C (235°F) |
| Odor | Phenol-like |
| Assay Percent Range | 99% |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Beilstein | 1817334 |
| Merck Index | 14,1585 |
| Show More |
Safety and Handling
| Hazard Category | H315-H318-H361f |
| Hazard Statement | GHS H Statement H318-H317-H335-H315 Causes serious eye damage. May cause an allergic skin reaction. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 202-679-0 |
| RTECSNumber | SJ8925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.