Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylacetylene, 90+%

Catalog No. AA4392009
Click to view available options
Quantity:
2 g
10 g

CAS: 772-38-3 | C12H14 | 158.244 g/mol

4-tert-Butylphenylacetylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-38-3
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00190197
InChI Key ZSYQVVKVKBVHIL-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
PubChem CID 2757302
IUPAC Name 1-tert-butyl-4-ethynylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C#C

Specifications

Density 0.878 g/mL
Boiling Point 130°C (40 mmHg)
Flash Point 82°C (180°F)
Odor Characteristic
Refractive Index 1.5297
Quantity 10 g
UN Number UN3082
Solubility Information Difficult to mix with water.
Formula Weight 158.24
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenylacetylene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.