Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B225125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2251-25G 25 g
B22515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2251-25G Supplier TCI America Supplier No. B225125G
Add to Cart
Add to Cart

Specifications

CAS 123324-71-0
Melting Point 184°C
Molecular Formula C10H15BO2
Quantity 25 g
MDL Number MFCD01009697
Synonym 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid
InChI Key MNJYZNVROSZZQC-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(C)(C)C)(O)O
IUPAC Name (4-tert-butylphenyl)boronic acid
Molecular Weight (g/mol) 178.038
PubChem CID 2734320
Formula Weight 178.04
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.