Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-tert-Butylpropiophenone 95.0+%, TCI America™
SDP

Catalog No. B16091ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16091ML 1 mL
B16095ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16091ML Supplier TCI America Supplier No. B16091ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71209-71-7
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.286
MDL Number MFCD00100434
InChI Key AQNYEAINONORRY-UHFFFAOYSA-N
Synonym 4-tert-Butylpropionylbenzene
PubChem CID 595281
IUPAC Name 1-(4-tert-butylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Color Yellow
Boiling Point 162°C
Quantity 1 mL
UN Number 1993
Formula Weight 190.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-tert-Butylpropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.