Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Thien-2-yltetrahydropyran-4-yl)methyl toluene-4-sulfonate, 97%, Thermo Scientific™

Catalog No. CC62244CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62244CB 250 mg Amber Glass Bottle
CC62244DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62244CB Supplier Thermo Scientific Chemicals Supplier No. CC62244CB
Add to Cart

Chemical Identifiers

CAS 921938-87-6
Molecular Formula C17H20O4S2
Molecular Weight (g/mol) 352.463
MDL Number MFCD09879933
InChI Key OZWQPINOYPXHPX-UHFFFAOYSA-N
Synonym 4-thien-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulphonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzene-1-sulfonate, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulfonate, 4-thiophen-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate
PubChem CID 24229670
IUPAC Name (4-thiophen-2-yloxan-4-yl)methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2(CCOCC2)C3=CC=CS3

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 352.47
Percent Purity 97%
Chemical Name or Material (4-Thien-2-yltetrahydropyran-4-yl)methyl toluene-4-sulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.