Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Amylcyclohexyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. A182825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A182825G 25 g
A18285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A182825G Supplier TCI America Supplier No. A182825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61204-01-1
Molecular Formula C18H25N
Molecular Weight (g/mol) 255.41
MDL Number MFCD00075548
InChI Key FURZYCFZFBYJBT-UHFFFAOYSA-N
Synonym trans-4-4-pentylcyclohexyl benzonitrile, 4-trans-4'-n-pentylcyclohexyl benzonitrile, 4-trans-4-pentylcyclohexyl benzonitrile, 4-4-pentylcyclohexyl benzonitrile, 4-trans-4-pentylcyclohexyl-benzonitrile, 4-trans-4-n-pentylcyclohexyl benzonitrile, benzonitrile, 4-trans-4-pentylcyclohexyl, 4-trans-4-amylcyclohexyl benzonitrile, benzonitrile, 4-4-pentylcyclohexyl, 1-trans-4-amylcyclohexyl-4-cyanobenzene
PubChem CID 109063
IUPAC Name 4-(4-pentylcyclohexyl)benzonitrile
SMILES CCCCCC1CCC(CC1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 30°C
Color White-Yellow
Quantity 25 g
Formula Weight 255.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Amylcyclohexyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.