Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Tributylstannyl)-1-tritylimidazole

Catalog No. SYX001061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX001061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX001061G Supplier Sigma Aldrich Supplier No. SYX001061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds
  • It was also used as a substrate in the reaction of a series of alkynes with mercury(ii)acetate

Specifications

CAS 208934-35-4
Molecular Formula C34H44N2Sn
Linear Formula C34H44N2Sn
MDL Number MFCD10699150
Quantity 1 g
Molecular Weight (g/mol) 599.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.