Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)benzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T24535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24535G Supplier TCI America Supplier No. T24535G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65796-00-1
Molecular Formula C8H6ClF3O
Molecular Weight (g/mol) 210.58
MDL Number MFCD00052326
InChI Key LBMKFQMJURUPKC-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl chloride, 1-chloromethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxy benzylchloride, 4-trifluoromethoxybenzyl chloride, benzene, 1-chloromethyl-4-trifluoromethoxy, 4-chloromethyl phenoxy trifluoromethane, pubchem5804, acmc-209ns9, ksc623c1l, p-trifluoromethoxybenzyl chloride
PubChem CID 2777261
IUPAC Name 1-(chloromethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CCl)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 3265
Formula Weight 210.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.