Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T178825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1788-25G 25 g
T17881G 1 g
T17885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1788-25G Supplier TCI America Supplier No. T178825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 25 g
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid
SMILES B(C1=CC=C(C=C1)C(F)(F)F)(O)O
Molecular Weight (g/mol) 189.928
Formula Weight 189.93
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.