Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinyl-1,3-dioxolan-2-one 98.0+%, TCI America™
SDP

Catalog No. V0114100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0114100G 100 g
V011425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0114100G Supplier TCI America Supplier No. V0114100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4427-96-7
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00143315
InChI Key BJWMSGRKJIOCNR-UHFFFAOYSA-N
Synonym 4-vinyl-1,3-dioxolan-2-one, vinylethylene carbonate, 4-vinyl-1,3-dioxolan-2-one, stabilized with hydroquinone, 1,3-dioxolan-2-one, 4-ethenyl, acmc-1ajgp, vec;, ksc383e5n, 2-oxo-4-vinyl-1,3-dioxolane, 4-vinyl-1,3 dioxolan-2-one, 4-vinyl-1,3-dioxaolane-2-one
PubChem CID 2735038
IUPAC Name 4-ethenyl-1,3-dioxolan-2-one
SMILES C=CC1COC(=O)O1

Specifications

Color Colorless
Boiling Point 122°C
Quantity 100 g
UN Number 2810
Formula Weight 114.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinyl-1,3-dioxolan-2-one
EINECSNumber (5)-6745
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.