Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinyl-1-cyclohexene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018409 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8409 25 mL
11-101-9952 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8409 Supplier MilliporeSigma Supelco Supplier No. 9495025ML
Add to Cart
Add to Cart

4-Vinyl-1-cyclohexene can be synthesized via cyclo-dimerization of butadiene in the presence of nitrosyl iron catalysts like [Fe(NO)2(CO)2] or [{FeCl(NO)2}2]/reductant.

Featured Industries: Environmental and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-40-3
Ignition Point 517°F
Vapor Pressure 10.2 mmHg (25°C)
Vapor Density 3.76 (vs. air)
Density 0.831 g/mL (at 20°C)
Flash Point 16°C
Physical Form Neat
Linear Formula C6H9 CH=CH2
MDL Number MFCD00001576
Melting Point -101°C
Boiling Point 126°C to 127°C
Synonym 4-Ethenyl-1-cyclohexene; NSC 15760
Type Analytical Standard
Quantity 25 mL
Formula Weight 108.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.