Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinyl-1-cyclohexene, 97%, stabilized

Catalog No. AC140880050
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 100-40-3 | C8H12 | 108.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-40-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001576
InChI Key BBDKZWKEPDTENS-UHFFFAOYSA-N
Synonym 4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3
PubChem CID 7499
ChEBI CHEBI:82377
IUPAC Name 4-ethenylcyclohexene
SMILES C=CC1CCC=CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101.0°C
Color Beige-Brown to White
Density 0.8320g/mL
Boiling Point 126.0°C to 127.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4625 to 1.4645
Linear Formula C6H9CH=CH2
Quantity 5 mL
Beilstein 05, I, 63
Specific Gravity 0.832
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in benzene and ether
Stabilizer 25 to 200ppm BHT
Viscosity 0.7 mPa.s (20°C)
Formula Weight 108.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Vinyl-1-cyclohexene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENT
EINECSNumber 202-848-9
RTECSNumber GW6650000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.