Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinyl-1-cyclohexene (stabilized with BHT) 95.0+%, TCI America™
SDP

Catalog No. V0023100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0023100ML 100 mL
V002325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0023100ML Supplier TCI America Supplier No. V0023100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-40-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00001576
InChI Key BBDKZWKEPDTENS-UHFFFAOYSA-N
Synonym 4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3
PubChem CID 7499
ChEBI CHEBI:82377
IUPAC Name 4-ethenylcyclohexene
SMILES C=CC1CCC=CC1

Specifications

Color Yellow
Boiling Point 128°C
Quantity 100 mL
UN Number 3295
Formula Weight 108.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinyl-1-cyclohexene (stabilized with BHT)
EINECSNumber (3)-2229
RTECSNumber GW6650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.