Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylbenzyl chloride, 90%, technical

Catalog No. AC422521000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422521000 100 g Glass bottle
AC422520250 25 g Glass bottle
AC422525000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC422521000 Supplier Thermo Scientific Chemicals Supplier No. 422521000
Add to Cart

CAS: 1592-20-7 | C9H9Cl | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-20-7
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00051362
InChI Key ZRZHXNCATOYMJH-UHFFFAOYSA-N
Synonym 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0
PubChem CID 74126
IUPAC Name 1-(chloromethyl)-4-ethenylbenzene
SMILES ClCC1=CC=C(C=C)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 1.0800g/mL
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5731 to 1.5751
Linear Formula CH2CHC6H4CH2Cl
Quantity 100 g
Specific Gravity 1.08
Stabilizer about 500ppm 4-tert-Butylcatechol, about 500ppm Nitrocresol + Nitrophenol
Formula Weight 152.62
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Vinylbenzyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 216-471-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.