Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-Vinylphenol solution 10100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W373923100GK
Encompass
4-Vinylphenol SOLUTIONon has been used as an HPLC internal standard during the identification of 4-vinyl derivatives formed via decarboxylation of hydroxycinnamic acids extracted from corn cobs using phenolic acid decarboxylase (PAD) enzyme extracted from Lactobacillus plantarum.
Catalog No. 50-290-4128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.