Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance Arc Flash Coat
SDP
Catalog No. 17001246 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-246 Medium
17-001-245 Small
17-001-247 Large
17-001-248 X-Large
17-001-249 2X-Large
17-001-250 3X-Large
17-001-251 4X-Large
17-001-252 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-246 Supplier National Safety Apparel™ Supplier No. C04UQUQ40MD32
Add to Cart
Add to Cart
For enhanced protection Westex™ UltraSoft is the ideal choice for superior comfort, improved safety, and above average durability.
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Breathable FR fabric
  • FR fabric will not melt, drip, or ignite
  • Generous cut allows for unrestrictive movement
  • FR hook and loop closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Available in a convenient kit
  • Made in the USA
Specifications
Closure Type FR Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size Medium
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269