Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 40 cal Arc Flash Kit (10)
SDP

Catalog No. 170011629 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1629 Medium
17-001-1624 Small
17-001-1634 Large
17-001-1639 X-Large
17-001-1644 2X-Large
17-001-1649 3X-Large
17-001-1654 4X-Large
17-001-1659 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1629 missing translation for 'mfr' National Safety Apparel™ missing translation for 'supplierNo' KIT4SC40MD10
Add to Cart
Add to Cart

Kit Includes: 40 cal Compliance short coat & bib overall, Arc flash hood with PureView™ faceshield and hard hat, safety glasses, hood bag and gear bag

  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • FR cuffs improve fit with electrical gloves
  • PureView™ faceshield technology features a greater range of visibility
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Material Cotton, Nylon
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 40 cal/cm²
Suspender Adjustable
Closure Type FR Hook and Loop
Color Navy
Size Medium
Seam Type Stitched
Ankle Style Open
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.