Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ RevoLite™ 40 cal Arc Flash Kit with Lift Front Hood and Voltage Glove (09)
SDP

Catalog No. 170011907 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1907 2X-Large
17-001-1887 Small
17-001-1892 Medium
17-001-1897 Large
17-001-1902 X-Large
17-001-1912 3X-Large
17-001-1917 4X-Large
17-001-1922 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1907 Supplier National Safety Apparel™ Supplier No. KIT4SCLT40LF2X09
Add to Cart
Add to Cart

50% lighter than the standard 40 cal kit

  • 50% Lighter than our standard 40 cal kits
  • FR Short Coat and Bib Overall
  • 14 in. Class 2 rubber voltage insulating gloves
  • 12 in. leather glove protectors
  • Lift front arc flash hood unit with PureView face shield and hard hat
  • Safety glasses
  • face shield unit bag
  • Glove bag
  • Gear bag

Specifications

Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material RevoLite™
Arc Flash Rating 40 cal/cm²
Size 2X-Large
Suspender Adjustable
Closure Type FR Hook and Loop Front Closure, Buckle
Ankle Style Open
Seam Type Stitched
Color Olive Green
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.