Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ RevoLite™ 40 cal Arc Flash Kit, No Gloves
SDP

Catalog No. 170011929 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1929 X-Large
17-001-1926 Small
17-001-1927 Medium
17-001-1928 Large
17-001-1930 2X-Large
17-001-1931 3X-Large
17-001-1932 4X-Large
17-001-1933 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1929 Supplier National Safety Apparel™ Supplier No. KIT4SCLT40NGXL
Add to Cart
Add to Cart

Arc flash kit with short coat and bib overall with no gloves

  • 50% lighter than our standard 40 cal kits
  • FR hook and loop closure with drawstring waist
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 40 cal/cm²
Suspender Adjustable
Ankle Style Open
Closure Type FR Hook and Loop
Material RevoLite™
Color Olive Green
Seam Type Stitched
Size X-Large
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.