Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ 40 Cal Side Fan Assembly For Revolite Hood
SDP

$296.00

Specifications

Size Medium
Material Plastic
Color Black
Seam Type Stitched
For Use With (Equipment) Arc Flash Hoods
View More Specs

Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
17-001-688
SDP
View Documents
National Safety Apparel™
FANNPLS
Left Side
Each for $296.00
Only null left
Add to Cart
 
17-001-690
SDP
View Documents
National Safety Apparel™
FANNPRS
Right Side
Each for $296.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Medium
Black
Arc Flash Hoods
Plastic
Stitched
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.