Learn More
Sino Biological Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP Protein (aa:Met1-Asn637, His Tag) 100ug
Supplier:  Sino Biological 40094V08B1100
Encompass_Preferred
A DNA sequence encoding Sudan ebolavirus (strain Uganda-00) GP (YP_138523.1) (Met1-Asn637) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.