Learn More
Sino Biological HIV-2 gp36 Protein (subtype CRF01_AB, strain 07JP_NMC716_clone_01) (His & MBP Tag) 100ug
Supplier:  Sino Biological 40112V10E100
Encompass_Preferred
A DNA sequence encoding the HIV-2 envelope glycoprotein gp160 (BAM76170.1)(Gln547-Tyr670), termed as gp36, was expressed with the N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-162-0376
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.