Learn More
Sino Biological HIV-1 gp120 Protein (group P, strain RBF168) (His Tag) 100ug
Supplier:  Sino Biological 40259V08H1100
Encompass_Preferred
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 (ACY40659.1) extracellular domain (Tyr38-Arg527), termed as gp120, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0674
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.