Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mmao-12

Catalog No. 404594800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-459-4800ML 800 mL
40-459-425ML 4 x 25 mL
40-459-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 40-459-4800ML Supplier Sigma Aldrich Supplier No. 404594800ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dialkylmetallocenes can undergo exchange reactions with methylaluminoxane.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 206451-54-9
Density 0.895 g/mL at 25°C
Boiling Point 111°C
Linear Formula [(CH3)0.95(n-C8)H17]0.05AlO]n
Quantity 800 mL
Synonym Methylaluminoxane solution; Modified methylaluminoxane (MAO) solution in toluene
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.