Learn More
Sino Biological Influenza B (B/Wisconsin/01/2012) Hemagglutinin / HA1 Protein (His Tag) 100ug
Supplier:  Sino Biological 40462V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Wisconsin/01/2012) hemagglutinin (translated amino acids of EPI354025) (Met1-Arg362), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0920
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.