Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodiisobutyloctadecylsilane

Catalog No. 4047135ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-471-35ML 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 40-471-35ML Supplier Sigma Aldrich Supplier No. 4047135ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

remainder octadecane

Specifications

CAS 28539-02-8
Melting Point 150°C to 152°C
Molecular Formula C7H7N3O
Linear Formula C7H7N3O
Refractive Index n20/D 1.459
MDL Number MFCD00179118
Quantity 50 g
Synonym 1-(Hydroxymethyl)benzotriazole; 1-Benzotriazolemethanol; NSC 12463
Molecular Weight (g/mol) 149.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.