Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methylimidazolium Chloride

Catalog No. 404771KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-477-1KGF 1 kg
40-477-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 40-477-1KGF Supplier Sigma Aldrich Supplier No. 404771KGF
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methylimidazolium chloride behaves as a solvent and catalyst during the formation of 5-hydroxymethylfurfural from fructose and sucrose via acid-catalyzed dehydration.

Specifications

CAS 35487-17-3
Molecular Formula C4H6N2 -+ HCl
Linear Formula C4H6N2 -+ HCl
MDL Number MFCD06798176
Quantity 1 kg
Synonym 1-Methylimidazole hydrochloride
Molecular Weight (g/mol) 118.56 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.