Learn More
Sino Biological Coxsackievirus A16 (Cox A16) (strain G-10) VP1 Protein (Fc Tag)
Supplier:  Sino Biological 40535V02B100
Encompass_Preferred
A DNA sequence encoding the VP1 of Coxsackievirus A16 (Cox A16) (strain G-10) (AAA50478.1) (Gly568-Lys862) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1072
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.