Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 406 Double Latex-Coated Insulated Cold-Weather Gloves

Catalog No. 19395350 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-395-350 2X-Large
19-395-347 Medium
19-395-348 Large
19-395-349 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-395-350 Supplier SHOWA™ Supplier No. 406XXL10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water-repellant gloves are designed to remain dry when used with liquids, while intelligent acrylic insulation protects hands from freezing temperatures down to -30°C.

  • Double-dipped latex coating with a rough texture for enhanced grip capabilities
  • Intelligent acrylic insulation protects hands from temperatures down to -30°C
  • Insulation helps to reduce perspiration inside the glove to preserve grip and tactility
  • High visibility orange exterior increases user safety in low-light applications
  • Seamless knit construction for a snug fit to increase warmth and dexterity

Specifications

ANSI Abrasion Rating ANSI 2
Coated Area Palm
Construction Seamless Knit
Coating Material Latex
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A1
Exterior Finish Partially Textured
Hand Specific
Latex Free No
Material Nylon, Polyester
Size 2X-Large
For Use With (Application) Brick Laying, Outdoor Maintenance, Refrigeration, and Shipping/Receiving
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Liner Material Acrylic
Puncture Rating ANSI/ISEA 105-16 Level 1
Series 406
Thumb Style Straight
Length (English) 11 in.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.