Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl pyrocarbonate

Catalog No. 40718100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
250 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-718-100ML 100 mL
40-718-250ML 250 mL
40-718-25ML 25 mL
40-718-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 40-718-100ML Supplier Sigma Aldrich Supplier No. 40718100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Diethyl pyrocarbonate (dep) is a bactericidal agent and decomposes in water to co2 and ethanol
  • Dep inhibits enzymes such as dnase and rnase without affecting nucleic acids
  • It is useful for the preparation of mrna from tissues.modification reagent for his and tyr residues in proteins
  • Robust probe for structural disruptions in dsdna, reacting with fully or partially unstacked bases.inactivates rnase in solution at about 0.1% (v/v), thus protecting RNA against degradation

Specifications

CAS 1609-47-8
Density 1.121 g/mL at 20°C, 1.101 g/mL (at 25°C (literature))
Boiling Point 93°C to 94°C (18 mmHg)
Molecular Formula C6H10O5
Refractive Index n20/D 1.398 (literature); n20/D 1.398
Linear Formula O(COOC2H5)2
MDL Number MFCD00009106
Quantity 100 mL
Synonym DEP; DEPC; Diethyl dicarbonate; Diethyl oxydiformate; Ethoxyformic acid anhydride
Molecular Weight (g/mol) 162.14
Percent Purity 99% (NT)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.