Learn More
Description
- 3-methoxy-3-methyl-1-butanol is widely used as solvent for paints, inks, and fragrances
- Kinetics of the reaction of 3-methoxy-3-methyl-1-butanol with oh radicals using a relative rate method has been reported
- It has been identified as oxidation products of dyes such as methyl yellow, methyl red and methyl orange.3-methoxy-3-methyl-1-butanol may be employed as raw material for the production of industrial detergents
Specifications
Specifications
| CAS | 56539-66-3 |
| Density | 0.926 g/mL (at 20°C (literature)) |
| Boiling Point | 173°C to 175°C (lit.) |
| Molecular Formula | C6H14O2 |
| Refractive Index | n20/D 1.428 (literature) |
| Linear Formula | (CH3)2 C(OCH3)CH2CH2OH |
| Quantity | 1 L |
| MDL Number | MFCD00044771 |
| Molecular Weight (g/mol) | 118.17 |
| Percent Purity | ≥98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.