Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(diallyldimethylammonium chloride) solution

Catalog No. 4090144L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-901-44L 4 L
40-901-41L 1 L
40-901-425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 40-901-44L Supplier Sigma Aldrich Supplier No. 4090144L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(diallyldimethylammonium chloride) solution (PDADMAC) is a cationic polyelectrolyte that is widely utilized as a flocculant. It is prepared by the radical polymerization of diallyldimethylammonium chloride (DADMAC) in presence of a catalyst in the form of an organic peroxide.PDADMAC can be used as an electrolytic solution in combination with mesoporous silica to form a responsive carrier system for potential usage in drug delivery. It can be also be coated on gold nanorods to enhance the cellular uptake in biomedical systems. It is used in the surface modification of indium tin oxide (ITO) substrate which may be used in chemical sensors or biosensor based applications.

Specifications

CAS 26062-79-3
Density 1.04 g/mL (at 25°C)
Refractive Index n20/D 1.375
Linear Formula (C8H16ClN)n
Quantity 4 L
MDL Number MFCD00192409
Synonym PDADMAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.