Learn More
VEOLIA WTS Certified TOC Vials, 40-mL, 72/cs
Supplier: VEOLIA WTS HMI 90606
Pre-cleaned and certified (<10 ppb TOC) 40-mL glass vials with caps and Teflon-lined septa. These vials arethe right choice for strict regulatory compliance testing. 72/cs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.