Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 40nm Colloidal Gold Nanoparticles 1 OD
SDP

Catalog No. 2010100 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-101-00 100 mL
20-100-10 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only
Catalog No. 20-101-00 Supplier R&D Systems™ Supplier No. 2010100

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

For Research Use Only

A Novus product, part of R&D Systems.

  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development

Specifications

Target 40nm Colloidal Gold Nanoparticles 1 OD
Product Type Antibody Labeling Kit
Content And Storage Store at 4°C. Do not freeze.
Product Line Novus
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.