Learn More
Bulbman Inc SYLVANIA F40T12WX 40W 48IN
SDP
Supplier:  Bulbman Inc 120283
Encompass_Preferred
48", 40W, T12 rapid start fluorescent lamp, deluxe warm white phosphor, 3500K color temperature, 87 CRI. 20000 Avg rated life at 3 hrs/start, 28800 Avg rated life at 12 hrs/start
Catalog No. NC0927174
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.