Learn More
Abcam 410/450 nm Extracellular Vesicle Membrane Staining Kit, 500 Tests
Description
- Assay type: Cell-based
- Applications: Flow Cyt
- Detection method: Flow cytometry-fluorescent
- Sample types: Adherent cells, Suspension cells
- Reactive species: Mammals
- Results type: Quantitative
- Protocol: Labeling in < 4 hrs with 30 secs hands-on time
- Product efficiency: 100% antibody recovery
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Extracellular Vesicle Membrane |
| For Use With (Equipment) | Flow Cytometer |
| Format | Kit |
| No. of Reactions | 500 Tests |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Sample Type | Adherent cells, Suspension cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.