Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibenzyl N,N-diisopropylphosphoramidite

Catalog No. 41643625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-643-625ML 25 mL
41-643-65ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-643-625ML Supplier Sigma Aldrich Supplier No. 41643625ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dibenzyl n,n-diisopropylphosphoramidite may be used for the preparation of phosphopeptides
  • It may be used for the synthesis of a gdp (guanosine diphosphate) analog, sml-8-73-1

Specifications

CAS 108549-23-1
Density 1.028 g/mL (at 25°C (literature))
Boiling Point 130°C (0.55 mmHg)
Molecular Formula C20H28NO2P
Refractive Index n20/D 1.535 (literature)
Linear Formula [(CH3)2 CH]2 NP(OCH2C6H5)2
Quantity 25 mL
MDL Number MFCD00191988
Molecular Weight (g/mol) 345.42
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.