Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Isopropoxy-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

Catalog No. 4171495ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-714-95ML 5 mL
41-714-9500ML 500 mL
41-714-9100ML 100 mL
41-714-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 41-714-95ML Supplier Sigma Aldrich Supplier No. 4171495ML
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane can be used as a reagent to borylate arenes and to prepare fluorenylborolane.It can also be used in the synthesis of following intermediates for generating conjugated copolymers:9,9-Dioctyl-2,7-bis(4,4,5,5-tetramethyl1,3,2-dioxaborolane-2-yl)dibenzosilole.3,9-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,11-di(1-decylundecyl)indolo[3,2-b]carbazole.2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dioctylfluorene.2,7-Bis(4'sa,4'sa,5'sa,5'sa-tetramethyl-1'sa,3'sa,2'sa-dioxaborolan-2'sa-yl)-N-9'sa'sa-heptadecanylcarbazole.

Specifications

CAS 61676-62-8
Density 0.912 g/mL at 25°C
Boiling Point 73°C (15 mmHg, lit.)
Molecular Formula C9H19BO3
Linear Formula C9H19BO3
Refractive Index n20/D 1.409
MDL Number MFCD00192241
Quantity 5 mL
Molecular Weight (g/mol) 186.06 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.