Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(melamine-co-formaldehyde) methylated, solution

Catalog No. 418560250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-856-0250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-856-0250ML Supplier Sigma Aldrich Supplier No. 418560250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(melamine-co-formaldehyde) methylated, solution (PMF) is a heat initiated cross-linking agent that forms an optically transparent solution. It can be linked with poly(4-vinly phenol) (PVP) by attaching with phenol groups that reduce the hydroxyl units in the blend.PMF can be used in the preparation of PVP-based ink, which can be used in inkjet printing for the development of electronic components such as:organic thin film transistor radio frequency capacitors parallel plate capacitors metal insulated metal (MIM) capacitor

Specifications

CAS 68002-20-0
Density 1.157 g/mL (at 25°C)
Boiling Point 118°C
Linear Formula (C4H8N6O)n
Refractive Index n20/D 1.515
MDL Number MFCD00197912
Quantity 250 mL
Molecular Weight (g/mol) 432
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.