Learn More
Eisco™ 42 Piece Lab Glass Set
Catalog No. 04999134 Shop All Eisco Products
Change view
Click to view available options
Quantity:
42 Pieces
Catalog No. Quantity
04-999-134 42 Pieces
1 options
Catalog No. 04-999-134 Supplier Eisco™ Supplier No. CH202409
Add to Cart
Add to Cart
42 Piece Lab Set Kit containing Low Form Beakers, Graduated Cylinders, Reagent Bottles and Conical Flasks.
  • 3 x 50 mL Low Form Beaker, with spout made of borosilicate glass, graduated.
  • 3 x 100 mL Low Form Beaker, with spout made of borosilicate glass, graduated.
  • 6 x 250 mL Low Form Beaker, with spout made of borosilicate glass, graduated,
  • 2 x 100 mL Reagent Bottle, graduated, borosilicate glass, with screw cap (GL 45).
  • 3 x 250 mL Reagent Bottle, graduated, borosilicate glass, with screw cap (GL 45).
  • 3 x 500 mL Reagent Bottle, graduated, borosilicate glass, with screw cap (GL 45).
  • 2 x 1000 mL Reagent Bottle, graduated, borosilicate glass, with screw cap (GL 45).
  • 1 x 100 mL Graduated Cylinder, class 'B', Hex. base with spout, borosilicate glass, Blue Graduation with plastic guard.
  • 1 x 250 mL Graduated Cylinder, class 'B', Hex. base with spout, borosilicate glass, Blue Graduation with plastic guard.
  • 1 x 500 mL Graduated Cylinder, class 'B', Hex. base with spout, borosilicate glass, Blue Graduation with plastic guard.
  • 3 x 250 mL Conical Flask with wide neck, borosilicate glass
  • 6 x 500 mL Conical Flask with wide neck, borosilicate glass
  • 3 x 1000 mL Conical Flask with wide neck, borosilicate glass
  • 2 x 400 mL Low Form Beaker with spout made of borosilicate glass, graduated.
  • 2 x 600 mL Low Form Beaker with spout made of borosilicate glass, graduated.
  • 1 x 1000 mL Low Form Beaker with spout made of borosilicate glass, graduated.
Specifications
Material Glass
Quantity 42 Pieces
Product Type Chemistry Kit