Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ludox™ Tm-50 Colloidal Silica

Catalog No. 4207784L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-077-84L 4 L
42-077-818L 18 L
42-077-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 42-077-84L Supplier Sigma Aldrich Supplier No. 4207784L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

LUDOX™ TM-50 colloidal silica is an aqueous dispersion of silica particles with intermediate particle size and narrow particle size distribution.LUDOX™ TM-50 colloidal silica can be used:As a silica source to synthesize tetrapropylammonium(TPA)-silicate-1 colloidal particles.As a removable template to prepare mesoporous carbon materials by using a resorcinol-formaldehyde resin as a carbon precursor.As a separation layer to synthesize non-spherical flexible poly(butylacrylate) (PBA) polymer particles by concentrated emulsion polymerization.LUDOX is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 7631-86-9
Density 1.4 g/mL at 25°C
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232
Quantity 4 L
Synonym Silica preparation
Molecular Weight (g/mol) 60.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.