Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Tert-Butyl-3-Ethylcarbodiimide

Catalog No. 4263935G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-639-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-639-35G Supplier Sigma Aldrich Supplier No. 4263935G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for: Guanylation of aminesSynthesis of tantalum dimethylamido diamidato and guanidinato complexes for CVD of nitride thin filmsPreparation of titanium amidinate guanidinate alkylamide complex precursors for HfO2 filmsKetene cycloadditionsHydroamination of carbodiimides using group-2-element amides as precatalystsSynthesis of cobalt amidinato derivative complexes for studies involving thermal stability, vapro pressure, vaporization rate and thermal decomposition properties

Specifications

CAS 13601-08-6
Density 1.03 g/mL at 25°C
Refractive Index n20/D 1.432
MDL Number MFCD00801255
Quantity 5 g
Synonym Nalpha-Fmoc-N(im)-trityl-L-histidine 4-benzyloxybenzyl ester polymer-bound; PS-PHB-His(Trt)-Fmoc
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.