Learn More
Burch Bottle & Packaging Inc 43-400 WHITE POLYPROPYLENE P/E
Supplier:  Burch Bottle & Packaging Inc 2WP0432PX
Encompass
Stock number 2WP0432PX is a 43-400 White Polypropylene injection molded closure. This cap features ribbed sides allowing for easy application/removal and a matte top. The F217 (foam) liner inside is a general liner that is compatible with a variety of products and reduces the transmission of moisture.
Catalog No. NC1778424
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.