Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane triglycidyl ether

Catalog No. 430269250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-026-9250ML 250 mL
43-026-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-026-9250ML Supplier Sigma Aldrich Supplier No. 430269250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylolpropane triglycidyl ether (TMPTE) has a trifunctional aliphatic glycidyl ether epoxide monomer mainly used as a crosslinking agent which provides chemical and mechanical resistance.TMPTE is used in the synthesis of azidated trimethylolpropane triglycidyl ether for the preparation of polyether polyol based hyperbranched polyurethanes. It can be coated on polycarbonated chips for improving chemical resistance from different organic solvents. Cross-linking of TMPTE can be done with poly(4-venylphenol) (PVP) for the preparation of organic field effect transistors (OFETs).

Specifications

CAS 30499-70-8
Density 1.157 g/mL (at 25°C)
Molecular Formula C15H26O6
Refractive Index n20/D 1.477 (literature)
Linear Formula C15H26O6
MDL Number MFCD00274208
Quantity 250 mL
Synonym 1,1,1-Trimethylolpropane triglycidyl ether; 2,2 -(((2-Ethyl-2-((oxiran-2-ylmethoxy)methyl)propane-1,3-diyl)bis(oxy))bis(methylene))bis(oxirane); Triglycidyl trimethylolpropane ether
Molecular Weight (g/mol) 302.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.