Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Allyl-3-Methylimidazolium Chloride

Catalog No. 4396150GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-961-50GF 50 g
43-961-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-961-50GF Supplier Sigma Aldrich Supplier No. 4396150GF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.AMIM-Cl may be used: As a solvent to dissolve cellulose during the preparation of regenerated cellulose materials.As a solvent for the acetylation of cornhusk cellulose to form cornhusk cellulose acetates (CCAs).As a plasticizer for cornstarch to form AMIM-Cl-plasticized starch film.

Specifications

CAS 65039-10-3
Molecular Formula C7H11ClN2
Linear Formula C7H11ClN2
Quantity 50 g
MDL Number MFCD07784450
Synonym AMIM-Cl
Molecular Weight (g/mol) 158.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.