Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane

Catalog No. 441775100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-177-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-177-5100G Supplier Sigma Aldrich Supplier No. 441775100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane

Specifications

CAS 3806-34-6
Melting Point 44°C to 47°C
Molecular Formula C41H82O6P2
Linear Formula C41H82O6P2
MDL Number MFCD00071522
Quantity 100 g
Synonym Cyclic neopentanetetraylbis(octadecyl phosphite); Dioctadecyl pentaerythritol diphosphite; Distearyl pentaerythritol diphosphite; Phosphorous acid, dicyclic ester with pentaerythritol, dioctadecyl ester
Molecular Weight (g/mol) 733.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.