Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cesium Carbonate

Catalog No. 441902250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
250 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-190-2250G 250 g
44-190-25G 5 g
44-190-250G 50 g
44-190-2100G 100 g
44-190-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 44-190-2250G Supplier Sigma Aldrich Supplier No. 441902250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cesium Carbonate

Specifications

CAS 534-17-8
Melting Point 610°C
Molecular Formula CCs2O3
Linear Formula Cs2CO3
MDL Number MFCD00010957
Quantity 250 g
Synonym Carbonic acid dicesium
Molecular Weight (g/mol) 325.82 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.